4-Chloro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2601-1G
C2601-5G
TCC2601-1GEA
46.29
CAD
TCC2601-1G
TCC2601-5G
4-Chloro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-(trifluoromethyl)phenylboronic acid
CAS Number: 313545-41-4
MDL Number: MFCD04973086
Molecular Formula: C7H5BClF3O2
Molecular Weight: 224.37
Form: Crystal
Color: White
Melting point (°C): 182
Other Product Names / Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic Acid
Formula:
C₇H₅BClF₃O₂ Melting Pt: 182 °C |
MDL Number:
MFCD04973086 CAS Number: 313545-41-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...