Cefsulodin sodium salt hydrate ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Cefsulodin sodium, Sulcephalosporin

C2598-1G C2598-200MG
TCC2598-1GEA 480.48 CAD
TCC2598-1G TCC2598-200MG
Cefsulodin sodium salt hydrate ≥95.0% (by HPLC, total nitrogen)
Cefsulodin sodium salt hydrate

CAS Number: 1426397-23-0
Molecular Formula: C₂₂H₁₉N₄NaO₈S₂·xH₂O
Molecular Weight: 554.52
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Specific rotation [a]20/D: 19 deg (C=1, H2O)
Storage Temperature: 0-10°C

Formula: C22H19N4NaO8S2·xH2O
MW: 554.53 g/mol (anhy)
MDL Number: MFCD07793338
CAS Number: 52152-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d282077147f98e5a767e27ed84bdbe2a8ae89e5&vid=8d282077147f98e5a767e27ed84bdbe2a8ae89e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR