Caronic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2596-1G C2596-5G
TCC2596-1GEA 168.59 CAD
TCC2596-1G TCC2596-5G
Caronic anhydride ≥98.0% (by GC, titration analysis)
Caronic anhydride

CAS Number: 67911-21-1
MDL Number: MFCD09751198
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 55


Other Product Names / Synonyms: 3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

Formula: C₇H₈O₃
Boiling Pt: 137 °C
Melting Pt: 55 °C
MDL Number: MFCD09751198
CAS Number: 67911-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d146bffc2a649b75e45e28e700952619b518d916&vid=d146bffc2a649b75e45e28e700952619b518d916&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR