2-Chloro-6-methyl-3-nitropyridine ≥98.0%

Supplier: TCI America

Synonyms: 6-Chloro-2-methyl-5-nitropyridine, 6-Chloro-5-nitro-2-picoline, 2-Chloro-3-nitro-6-picoline, 2-Chloro-3-nitro-6-methylpyridine

C2594-1G C2594-25G C2594-5G
TCC2594-1GEA 50.56 CAD
TCC2594-1G TCC2594-25G TCC2594-5G
2-Chloro-6-methyl-3-nitropyridine ≥98.0%
2-Chloro-6-methyl-3-nitropyridine

CAS Number: 56057-19-3
MDL Number: MFCD03085820
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72


Other Product Names / Synonyms: 6-Chloro-5-nitro-2-picoline

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 72 °C
MDL Number: MFCD03085820
CAS Number: 56057-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90642fd44b44a0fb75861a64eedaae7b44a04b3e&vid=90642fd44b44a0fb75861a64eedaae7b44a04b3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR