2-Chloro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2-Chloro-5-(trifluoromethyl)phenylboronic Acid, Dihydroxy(2-chloro-5-(trifluoromethyl)phenyl)borane, (2-Chloro-5-(trifluoromethyl)phenyl)metaboric acid, 2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS Number: 182344-18-9
MDL Number: MFCD00797335
Molecular Formula: C7H5BClF3O2
Molecular Weight: 224.37
Form: Crystal
Color: White
Melting point (°C): 108
Other Product Names / Synonyms: 2-Chloro-5-(trifluoromethyl)benzeneboronic Acid
Formula:
C₇H₅BClF₃O₂ MW: 224.37 Melting Pt: 108 °C |
MDL Number:
MFCD00797335 CAS Number: 182344-18-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...