(2-Chloro-3-pyridinyl)methanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol

C2590-5G C2590-1G
TCC2590-5GEA 133.83 CAD
TCC2590-5G TCC2590-1G
(2-Chloro-3-pyridinyl)methanol ≥98.0%
(2-Chloro-3-pyridinyl)methanol

CAS Number: 42330-59-6
MDL Number: MFCD01935961
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 63


Other Product Names / Synonyms: 2-Chloro-3-(hydroxymethyl)pyridine

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 112 °C
Melting Pt: 63 °C
MDL Number: MFCD01935961
CAS Number: 42330-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b1c84ba38022ce19dc561b5ab6f2316fe9bf4a2&vid=a93ff241d90935830450dcc7afa2020b24abab64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR