2-Chloro-4-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid

C2589-1G C2589-5G
TCC2589-1GEA 66.36 CAD
TCC2589-1G TCC2589-5G
2-Chloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-pyridineboronic acid

CAS Number: 458532-96-2
MDL Number: MFCD03788416
Molecular Formula: C5H5BClNO2
Molecular Weight: 157.36
Form: Crystal
Color: White
Melting point (°C): 210
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Chloro-4-pyridylboronic Acid

Formula: C₅H₅BClNO₂
MW: 157.36
Melting Pt: 210 °C
MDL Number: MFCD03788416
CAS Number: 458532-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a70b8cc2e4f44b84818c882a551fd693da5a39&vid=88a70b8cc2e4f44b84818c882a551fd693da5a39&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR