4-Chlorobenzoylacetonitrile ≥98.0% (by GC)
Supplier: TCI America
C2582-25G
C2582-5G
TCC2582-25GEA
2414.24
CAD
TCC2582-25G
TCC2582-5G
4-Chlorobenzoylacetonitrile ≥98.0% (by GC)
4-Chlorobenzoylacetonitrile
CAS Number: 4640-66-8
MDL Number: MFCD00051625
Molecular Formula: C9H6ClNO
Molecular Weight: 179.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128
Other Product Names / Synonyms: 4'-Chloro-2-cyanoacetophenone
Formula:
C₉H₆ClNO Melting Pt: 128 °C |
MDL Number:
MFCD00051625 CAS Number: 4640-66-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...