2-Chlorobenzoylacetonitrile ≥97.0% (by GC)

Supplier: TCI America

C2580-25G C2580-5G
TCC2580-25GEA 876.9 CAD
TCC2580-25G TCC2580-5G
2-Chlorobenzoylacetonitrile ≥97.0% (by GC)
2-Chlorobenzoylacetonitrile

CAS Number: 40018-25-5
MDL Number: MFCD00051624
Molecular Formula: C9H6ClNO
Molecular Weight: 179.60
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 58
Storage Temperature: >-20°C


Other Product Names / Synonyms: 2'-Chloro-2-cyanoacetophenone

Formula: C₉H₆ClNO
Melting Pt: 58 °C
MDL Number: MFCD00051624
CAS Number: 40018-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88fa617cfc23412e48677d3bfb2259193a4a4375&vid=88fa617cfc23412e48677d3bfb2259193a4a4375&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR