4-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2577-1G C2577-5G
TCC2577-1GEA 55.3 CAD
TCC2577-1G TCC2577-5G
4-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-methylphenylboronic acid

CAS Number: 209919-30-2
MDL Number: MFCD02683107
Molecular Formula: C7H8BClO2
Molecular Weight: 170.40
Form: Crystal
Color: White
Melting point (°C): 195


Other Product Names / Synonyms: 4-Chloro-2-methylbenzeneboronic Acid, 4-Chloro-o-tolylboronic Acid

Formula: C₇H₈BClO₂
Melting Pt: 195 °C
MDL Number: MFCD02683107
CAS Number: 209919-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff2743aaa7f01a7b76b2473675be1d853fe5ec3&vid=fff2743aaa7f01a7b76b2473675be1d853fe5ec3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR