4-Chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C2574-25G C2574-5G
TCC2574-25GEA 308.1 CAD
TCC2574-25G TCC2574-5G
4-Chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
4-Chloro-6,7-dimethoxyquinazoline

CAS Number: 13790-39-1
MDL Number: MFCD01570172
Molecular Formula: C10H9ClN2O2
Molecular Weight: 224.64
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 185

Formula: C₁₀H₉ClN₂O₂
Melting Pt: 185 °C
MDL Number: MFCD01570172
CAS Number: 13790-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58eee619645bf7e068881292fb144b01f46a2950&vid=58eee619645bf7e068881292fb144b01f46a2950&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR