4-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(4-chloro-3-(trifluoromethyl)phenyl)borane, 4-Chloro-3-(trifluoromethyl)benzeneboronic acid, (4-Chloro-3-(trifluoromethyl)phenyl)metaboric acid

C2571-1G C2571-5G
TCC2571-1GEA 262.75 CAD
TCC2571-1G TCC2571-5G
4-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-(trifluoromethyl)phenylboronic acid

CAS Number: 176976-42-4
MDL Number: MFCD03094999
Molecular Formula: C7H5BClF3O2
Molecular Weight: 224.37
Form: Crystal
Color: White
Boiling point (°C): 236


Other Product Names / Synonyms: 4-Chloro-3-(trifluoromethyl)benzeneboronic Acid

Formula: C₇H₅BClF₃O₂
MW: 224.37
Boiling Pt: 236 °C
MDL Number: MFCD03094999
CAS Number: 176976-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f02eda59d861263e383bfd47af13bdc8c3af8827&vid=f02eda59d861263e383bfd47af13bdc8c3af8827&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR