4-Carbamoylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2570-1G
C2570-5G
TCC2570-1GEA
126.4
CAD
TCC2570-1G
TCC2570-5G
4-Carbamoylphenylboronic acid (contains varying amounts of Anhydride)
4-Carbamoylphenylboronic acid
CAS Number: 123088-59-5
MDL Number: MFCD03411940
Molecular Formula: C7H8BNO3
Molecular Weight: 164.96
Form: Crystal
Color: White
Melting point (°C): 234
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-Carbamoylbenzeneboronic Acid
Formula:
C₇H₈BNO₃ Melting Pt: 234 °C |
MDL Number:
MFCD03411940 CAS Number: 123088-59-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...