(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid ≥98.0%

Supplier: TCI America

Synonyms: Z-L-2-phenylglycine, (S)-2-(((Benzyloxy)carbonyl)amino)-2-phenylacetic acid, N-Benzyloxycarbonyl-L-phenylglycine, Z-Phg-OH

C2567-1G C2567-5G
TCC2567-1GEA 34.76 CAD
TCC2567-1G TCC2567-5G
(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid ≥98.0%
(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid

CAS Number: 53990-33-3
MDL Number: MFCD00077033
Molecular Formula: C16H15NO4
Molecular Weight: 285.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 133
Specific rotation [a]20/D: 116 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Z-L-2-phenylglycine, Z-Phg-OH

Formula: C16H15NO4
MW: 285.3 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00077033
CAS Number: 53990-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70257b2ffefe43732719d21bf66217858300860f&vid=70257b2ffefe43732719d21bf66217858300860f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR