N-Carbobenzoxy-D-2-phenylglycine ≥98.0%
Supplier: TCI America
Synonyms:
(R)-2-(((Benzyloxy)carbonyl)amino)-2-phenylacetic acid, Z-D-Phg-OH, Z-D-2-phenylglycine, Cbz-D-Phg-OH
C2566-1G
C2566-5G
TCC2566-1GEA
50.56
CAD
TCC2566-1G
TCC2566-5G
N-Carbobenzoxy-D-2-phenylglycine ≥98.0%
N-Carbobenzoxy-D-2-phenylglycine
CAS Number: 17609-52-8
MDL Number: MFCD00021703
Molecular Formula: C16H15NO4
Molecular Weight: 285.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 131
Specific rotation [a]20/D: -116 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Z-D-2-phenylglycine, Z-D-Phg-OH
Formula:
C₁₆H₁₅NO₄ MW: 285.29 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD00021703 CAS Number: 17609-52-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...