Cilnidipine ≥98.0%

Supplier: TCI America

Synonyms: 3-Cinnamyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl Ester, 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

C2564-1G C2564-5G
TCC2564-1GEA 137.46 CAD
TCC2564-1G TCC2564-5G
Cilnidipine ≥98.0%
Cilnidipine

CAS Number: 132203-70-4
MDL Number: MFCD00865853
Molecular Formula: C27H28N2O7
Molecular Weight: 492.53
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 110


Product Categories: Bioscience, Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists

Formula: C₂₇H₂₈N₂O₇
MW: 492.53 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00865853
CAS Number: 132203-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38e8345ca955f91c684d51069bb0fa1bbd8fb973&vid=38e8345ca955f91c684d51069bb0fa1bbd8fb973&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR