N-Cbz-4-piperidinecarboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: 1-Benzyloxycarbonyl-4-piperidinecarboxylic acid, 1-Benzyloxycarbonylisonipecotic acid, 1-Carbobenzoxy-4-piperidinecarboxylic acid, 1-Cbz-isonipecotic acid, 1-Carbobenzoxyisonipecotic acid, 1-Cbz-4-piperidinecarboxylic acid, N-Cbz-4-Piperidinecarboxylic acid

C2563-1G C2563-25G C2563-5G
TCC2563-1GEA 67.94 CAD
TCC2563-1G TCC2563-25G TCC2563-5G
N-Cbz-4-piperidinecarboxylic acid ≥97.0%
N-Cbz-4-piperidinecarboxylic acid

CAS Number: 10314-98-4
MDL Number: MFCD01568759
Molecular Formula: C14H17NO4
Molecular Weight: 263.29
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 73


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Cbz-4-piperidinecarboxylic Acid, 1-Carbobenzoxyisonipecotic Acid, 1-Cbz-isonipecotic Acid

Formula: C14H17NO4
MW: 263.2900 g/mol
Melting Pt: 73 °C
MDL Number: MFCD01568759
CAS Number: 10314-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efcc66dffa16f99443ab83b49c7d019bab69bbaa&vid=efcc66dffa16f99443ab83b49c7d019bab69bbaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR