5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
C2549-1G
C2549-5G
TCC2549-1GEA
63.2
CAD
TCC2549-1G
TCC2549-5G
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by GC, titration analysis)
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid
CAS Number: 61727-33-1
MDL Number: MFCD00173907
Molecular Formula: C6H5ClN2O2S
Molecular Weight: 204.63
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 170
Formula:
C₆H₅ClN₂O₂S Melting Pt: 170 °C |
MDL Number:
MFCD00173907 CAS Number: 61727-33-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...