3-Chloro-4-formylpyridine ≥98.0% (by GC)

Supplier: TCI America

C2542-1G C2542-5G
TCC2542-1GEA 52.14 CAD
TCC2542-1G TCC2542-5G
3-Chloro-4-formylpyridine ≥98.0% (by GC)
3-Chloro-4-formylpyridine

CAS Number: 72990-37-5
MDL Number: MFCD06200712
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 93
Melting point (°C): 60


Other Product Names / Synonyms: 3-Chloroisonicotinaldehyde

Formula: C₆H₄ClNO
Boiling Pt: 93 °C
Melting Pt: 60 °C
MDL Number: MFCD06200712
CAS Number: 72990-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad13ace3c08e9648c69ac1999399cf96ba4542ef&vid=ad13ace3c08e9648c69ac1999399cf96ba4542ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR