2-Fluoronicotinonitrile ≥98.0% (by GC)

Supplier: TCI America

C2538-25G C2538-5G
TCC2538-25GEA 297.04 CAD
TCC2538-25G TCC2538-5G
2-Fluoronicotinonitrile ≥98.0% (by GC)
2-Fluoronicotinonitrile

CAS Number: 3939-13-7
MDL Number: MFCD03095082
Molecular Formula: C6H3FN2
Molecular Weight: 122.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 29


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoronicotinonitrile

Formula: C₆H₃FN₂
MDL Number: MFCD03095082
CAS Number: 3939-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5ceaefae1dc10a16d0a7d09b21add81a043b816&vid=d5ceaefae1dc10a16d0a7d09b21add81a043b816&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR