N-Boc-L-aspartic acid 4-cyclohexyl ester ≥98.0%

Supplier: TCI America

Synonyms: Boc-Asp(OcHex)-OH, 4-Cyclohexyl-N-(tert-butoxycarbonyl)-L-aspartate, Boc-Asp(OcHx)-OH, N-Boc-L-aspartic Acid 4-Cyclohexyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Cyclohexyl Ester, 4-Cyclohexyl N-Boc-L-aspartate

C2535-25G C2535-5G
TCC2535-25GEA 268.6 CAD
TCC2535-25G TCC2535-5G
N-Boc-L-aspartic acid 4-cyclohexyl ester ≥98.0%
N-Boc-L-aspartic acid 4-cyclohexyl ester

CAS Number: 73821-95-1
MDL Number: MFCD00061996
Molecular Formula: C15H25NO6
Molecular Weight: 315.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 96
Specific rotation [a]20/D: -22 deg (C=2, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Cyclohexyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Cyclohexyl Ester, N-Boc-L-aspartic Acid 4-Cyclohexyl Ester

Formula: C₁₅H₂₅NO₆
MW: 315.37 g/mol
Melting Pt: 96 °C
MDL Number: MFCD00061996
CAS Number: 73821-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1a1c52fcc9f9e6f7e2ac499bbc5fe8680cf654d&vid=a1a1c52fcc9f9e6f7e2ac499bbc5fe8680cf654d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR