2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloromethyl-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride, Lansoprazole Chloromethyl Impurity

C2532-25G C2532-5G
TCC2532-25GEA 131.14 CAD
TCC2532-25G TCC2532-5G
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride ≥98.0%
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride

CAS Number: 127337-60-4
MDL Number: MFCD00800224
Molecular Formula: C9H9ClF3NO
Molecular Weight: 276.08
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214
Storage Temperature: 0-10°C

Formula: C₉H₉ClF₃NO
MW: 276.08 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00800224
CAS Number: 127337-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62e2e38c72c9072cab1891258e0684c08323819e&vid=62e2e38c72c9072cab1891258e0684c08323819e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR