3-Chloro-4-(trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2528-25G C2528-5G
TCC2528-25GEA 211.72 CAD
TCC2528-25G TCC2528-5G
3-Chloro-4-(trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
3-Chloro-4-(trifluoromethoxy)aniline

CAS Number: 64628-73-5
MDL Number: MFCD00190126
Molecular Formula: C7H5ClF3NO
Molecular Weight: 211.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Freezing point (°C): 31

Formula: C₇H₅ClF₃NO
MDL Number: MFCD00190126
CAS Number: 64628-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75485df5882e97b4de22f497555be47c9ced935d&vid=75485df5882e97b4de22f497555be47c9ced935d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR