(4-Chlorophenyl)acetone ≥97.0% (by GC)

Supplier: TCI America

C2526-25G C2526-5G
TCC2526-25GEA 352.02 CAD
TCC2526-25G TCC2526-5G
(4-Chlorophenyl)acetone ≥97.0% (by GC)
(4-Chlorophenyl)acetone

CAS Number: 5586-88-9
MDL Number: MFCD00045214
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Freezing point (°C): 17
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 1-(4-Chlorophenyl)-2-propanone

Formula: C₉H₉ClO
Boiling Pt: 216 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00045214
CAS Number: 5586-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5322505b29041d70d00b6f6ec14d4959cc60fea5&vid=09b1eeacf8b8ba96c41e59aebabb15df317e7000&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR