3-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid

C2525-1G C2525-5G
TCC2525-1GEA 48.98 CAD
TCC2525-1G TCC2525-5G
3-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-methylphenylboronic acid

CAS Number: 175883-63-3
MDL Number: MFCD04039010
Molecular Formula: C7H8BClO2
Molecular Weight: 170.40
Form: Crystal
Color: White
Melting point (°C): 247


Other Product Names / Synonyms: 3-Chloro-4-methylbenzeneboronic Acid, 3-Chloro-p-tolylboronic Acid

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 247 °C
MDL Number: MFCD04039010
CAS Number: 175883-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c92c9353b83fa9570c91eb6ddd75fd836a27915e&vid=c92c9353b83fa9570c91eb6ddd75fd836a27915e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR