4-Carboxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2523-1G
C2523-5G
TCC2523-1GEA
197.5
CAD
TCC2523-1G
TCC2523-5G
4-Carboxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-3-chlorophenylboronic acid
CAS Number: 136496-72-5
MDL Number: MFCD06656268
Molecular Formula: C7H6BClO4
Molecular Weight: 200.38
Form: Crystal
Color: White
Melting point (°C): 232
Other Product Names / Synonyms: 4-Carboxy-3-chlorobenzeneboronic Acid
Formula:
C₇H₆BClO₄ Melting Pt: 232 °C |
MDL Number:
MFCD06656268 CAS Number: 136496-72-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...