Cromolyn disodium salt hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt, Disodium Cromoglycate
CAS Number: 15826-37-6
MDL Number: MFCD00057744
Molecular Formula: C23H16O11
Molecular Weight: 512.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: Disodium Cromoglycate, 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt
Formula:
C₂₃H₁₆O₁₁ .xH₂O MW: 512.33 g/mol (anhy) |
MDL Number:
MFCD00057744 CAS Number: 15826-37-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...