2-Chloro-5-formylpyridine ≥96.0%

Supplier: TCI America

Synonyms: 6-Chloronicotinaldehyde, 2-Chloropyridine-5-carboxaldehyde, 6-Chloropyridine-3-carboxaldehyde

C2519-1G C2519-5G
TCC2519-1GEA 71.1 CAD
TCC2519-1G TCC2519-5G
2-Chloro-5-formylpyridine ≥96.0%
2-Chloro-5-formylpyridine

CAS Number: 23100-12-1
MDL Number: MFCD03095223
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 6-Chloronicotinaldehyde, 2-Chloro-5-formylpyridine

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 79 °C
MDL Number: MFCD03095223
CAS Number: 23100-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dae67778edee45fac230dec8478f1028331f54d&vid=8dae67778edee45fac230dec8478f1028331f54d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR