3-Chloro-2,4-difluorobenzoic acid ≥98.0%

Supplier: TCI America

C2517-25G C2517-5G
TCC2517-25GEA 712.42 CAD
TCC2517-25G TCC2517-5G
3-Chloro-2,4-difluorobenzoic acid ≥98.0%
3-Chloro-2,4-difluorobenzoic acid

CAS Number: 154257-75-7
MDL Number: MFCD01631387
Molecular Formula: C7H3ClF2O2
Molecular Weight: 192.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 176

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Melting Pt: 176 °C
MDL Number: MFCD01631387
CAS Number: 154257-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb84c05cbadec6237af7025672af3b052be7d1ad&vid=fb84c05cbadec6237af7025672af3b052be7d1ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR