2-Chloro-4-hydroxypyridine ≥97.0%
Supplier: TCI America
C2516-1G
C2516-5G
TCC2516-1GEA
30.02
CAD
TCC2516-1G
TCC2516-5G
2-Chloro-4-hydroxypyridine ≥97.0%
2-Chloro-4-hydroxypyridine
CAS Number: 17368-12-6
MDL Number: MFCD01646108
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 169
Other Product Names / Synonyms: 2-Chloro-4-pyridinol
Formula:
C₅H₄ClNO MW: 129.55 Melting Pt: 169 °C |
MDL Number:
MFCD01646108 CAS Number: 17368-12-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...