5-Chloro-2-methylbenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-o-toluic acid

C2509-25G C2509-5G
TCC2509-25GEA 211.56 CAD
TCC2509-25G TCC2509-5G
5-Chloro-2-methylbenzoic acid ≥98.0%
5-Chloro-2-methylbenzoic acid

CAS Number: 7499-06-1
MDL Number: MFCD00045842
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 5-Chloro-o-toluic Acid

Formula: C₈H₇ClO₂
MW: 170.6
Melting Pt: 173 °C
MDL Number: MFCD00045842
CAS Number: 7499-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2655ca79d4997562b0f2cdfb6438411dbba110e2&vid=2655ca79d4997562b0f2cdfb6438411dbba110e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR