4-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2508-25G C2508-5G
TCC2508-25GEA 533.88 CAD
TCC2508-25G TCC2508-5G
4-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Chloro-o-toluic acid

CAS Number: 7499-07-2
MDL Number: MFCD00045852
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 4-Chloro-o-toluic Acid

Formula: C₈H₇ClO₂
Melting Pt: 173 °C
MDL Number: MFCD00045852
CAS Number: 7499-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d3060dcf804ec273d9fc342ea2d0f7eb271bdc7&vid=8d3060dcf804ec273d9fc342ea2d0f7eb271bdc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR