2-Chloro-5-nitropyrimidine ≥98.0% (by GC)

Supplier: TCI America

C2507-1G C2507-5G
TCC2507-1GEA 104.28 CAD
TCC2507-1G TCC2507-5G
2-Chloro-5-nitropyrimidine ≥98.0% (by GC)
2-Chloro-5-nitropyrimidine

CAS Number: 10320-42-0
MDL Number: MFCD04117995
Molecular Formula: C4H2ClN3O2
Molecular Weight: 159.53
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 109
Storage Temperature: <0°C

Formula: C₄H₂ClN₃O₂
Melting Pt: 109 °C
MDL Number: MFCD04117995
CAS Number: 10320-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f4e57b1626c0bc3a3550f686afb16666f36c2fa&vid=4f4e57b1626c0bc3a3550f686afb16666f36c2fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR