Z-Hyp-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2506-25G C2506-5G
TCC2506-25GEA 203.35 CAD
TCC2506-25G TCC2506-5G
Z-Hyp-OH ≥98.0% (by HPLC, titration analysis)
Z-Hyp-OH

CAS Number: 13504-85-3
MDL Number: MFCD00037329
Molecular Formula: C13H15NO5
Molecular Weight: 265.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 107
Specific rotation [a]20/D: -54 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: trans-N-Cbz-4-hydroxy-L-proline, Z-Hyp-OH

Formula: C13H15NO5
Melting Pt: 107 °C
MDL Number: MFCD00037329
CAS Number: 13504-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04bff529a006ae3cd1708f6a16d7d12929767d29&vid=04bff529a006ae3cd1708f6a16d7d12929767d29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR