2-Carboxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2501-1G C2501-5G
TCC2501-1GEA 79 CAD
TCC2501-1G TCC2501-5G
2-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
2-Carboxyphenylboronic acid

CAS Number: 149105-19-1
MDL Number: MFCD01318118
Molecular Formula: C7H7BO4
Molecular Weight: 165.94
Form: Crystal
Color: White
Melting point (°C): 162
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Carboxybenzeneboronic Acid

Formula: C₇H₇BO₄
Melting Pt: 162 °C
MDL Number: MFCD01318118
CAS Number: 149105-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff2e09df3df02acc65721d23c94fb7b41906f0c&vid=dff2e09df3df02acc65721d23c94fb7b41906f0c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR