Carnosic acid ≥97.0% (by HPLC)

Supplier: TCI America

C2488-100MG C2488-20MG
TCC2488-100MGEA 514.13 CAD
TCC2488-100MG TCC2488-20MG
Carnosic acid ≥97.0% (by HPLC)
Carnosic acid

CAS Number: 3650-09-7
MDL Number: MFCD02259459
Molecular Formula: C20H28O4
Molecular Weight: 332.44
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 190
Specific rotation [a]20/D: 184 deg (C=1, MeOH)
Storage Temperature: <0°C

Formula: C₂₀H₂₈O₄
Melting Pt: 190 °C
MDL Number: MFCD02259459
CAS Number: 3650-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e8fdaf1bbd99854e5fc6534925ff84c49df2477&vid=9e8fdaf1bbd99854e5fc6534925ff84c49df2477&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR