2-Chloro-3-formylpyridine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

C2487-1G C2487-5G
TCC2487-1GEA 33.18 CAD
TCC2487-1G TCC2487-5G
2-Chloro-3-formylpyridine ≥98.0% (by GC)
2-Chloro-3-formylpyridine

CAS Number: 36404-88-3
MDL Number: MFCD01315308
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 52


Other Product Names / Synonyms: 2-Chloro-3-formylpyridine, 2-Chloronicotinaldehyde

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 52 °C
MDL Number: MFCD01315308
CAS Number: 36404-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7231f17fb002b3d074d36638f7eb06cae24d6ae3&vid=7231f17fb002b3d074d36638f7eb06cae24d6ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR