5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥95.0%

Supplier: TCI America

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE

C2479-100MG C2479-20MG
TCC2479-100MGEA 1141.14 CAD
TCC2479-100MG TCC2479-20MG
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥95.0%
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C

Formula: C₂₅H₁₅NO₉
MDL Number: MFCD00077322
CAS Number: 92557-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8169eceabbb746c16f0e243c4458a75cbe28527c&vid=8169eceabbb746c16f0e243c4458a75cbe28527c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR