4-Chloro-3-fluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C2476-25G
C2476-5G
TCC2476-25GEA
541.94
CAD
TCC2476-25G
TCC2476-5G
4-Chloro-3-fluorophenol ≥98.0% (by GC, titration analysis)
4-Chloro-3-fluorophenol
CAS Number: 348-60-7
MDL Number: MFCD00042583
Molecular Formula: C6H4ClFO
Molecular Weight: 146.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 84
Melting point (°C): 54
Flash Point (°C): 113
Formula:
C₆H₄ClFO Boiling Pt: 84 °C Melting Pt: 54 °C Flash Pt: 113 °C |
MDL Number:
MFCD00042583 CAS Number: 348-60-7 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...