4-Chloro-7-azaindole ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2470-1G C2470-200MG
TCC2470-1GEA 163.69 CAD
TCC2470-1G TCC2470-200MG
4-Chloro-7-azaindole ≥97.0% (by GC, titration analysis)
4-Chloro-7-azaindole

CAS Number: 55052-28-3
MDL Number: MFCD08272232
Molecular Formula: C7H5ClN2
Molecular Weight: 152.58
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 179


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 4-Chloro-7-azaindole

Formula: C7H5ClN2
MW: 152.58 g/mol
Melting Pt: 179 °C
MDL Number: MFCD08272232
CAS Number: 55052-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f5127d11d2afc49a13cc89595788da375962a8c&vid=6f5127d11d2afc49a13cc89595788da375962a8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR