3-Chloro-p-toluic acid ≥98.0%
Supplier: TCI America
C2462-25G
C2462-5G
TCC2462-25GEA
219.62
CAD
TCC2462-25G
TCC2462-5G
3-Chloro-p-toluic acid ≥98.0%
3-Chloro-p-toluic acid
CAS Number: 5162-82-3
MDL Number: MFCD00045841
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 202
Other Product Names / Synonyms: 3-Chloro-p-toluic Acid
Formula:
C₈H₇ClO₂ Melting Pt: 202 °C |
MDL Number:
MFCD00045841 CAS Number: 5162-82-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...