5-Chloro-3-methylbenzo[b]thiophene ≥98.0% (by GC)

Supplier: TCI America

C2457-25G C2457-5G
TCC2457-25GEA 127.35 CAD
TCC2457-25G TCC2457-5G
5-Chloro-3-methylbenzo[b]thiophene ≥98.0% (by GC)
5-Chloro-3-methylbenzo[b]thiophene

CAS Number: 19404-18-3
MDL Number: MFCD00052506
Molecular Formula: C9H7ClS
Molecular Weight: 182.67
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 144
Melting point (°C): 35

Formula: C₉H₇ClS
MW: 182.67 g/mol
Boiling Pt: 144 °C
Melting Pt: 35 °C
MDL Number: MFCD00052506
CAS Number: 19404-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6642db32c85e7c5303f83a9eb7246852c400d42b&vid=6642db32c85e7c5303f83a9eb7246852c400d42b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR