8-Chloro-1-octanol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 8-Chloro-1-n-octanol

C2448-25ML C2448-5ML
TCC2448-25MLEA 187.23 CAD
TCC2448-25ML TCC2448-5ML
8-Chloro-1-octanol ≥97.0% (by GC)
8-Chloro-1-octanol

CAS Number: 23144-52-7
MDL Number: MFCD00040005
Molecular Formula: C8H17ClO
Molecular Weight: 164.67
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 113
Specific Gravity (20/20): 0.99

Formula: C₈H₁₇ClO
MW: 164.68 g/mol
Boiling Pt: 130 °C
Density: 0.99 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00040005
CAS Number: 23144-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bfa049b848fa522b52a7fbf2ec0e230c09e8555&vid=9d766fe4391d1a73f00c8b1b6e8e2f09ba77c1f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR