2-Chloro-3-fluoropyridine ≥98.0% (by GC)
Supplier: TCI America
C2447-25G
C2447-5G
TCC2447-25GEA
77.74
CAD
TCC2447-25G
TCC2447-5G
2-Chloro-3-fluoropyridine ≥98.0% (by GC)
2-Chloro-3-fluoropyridine
CAS Number: 17282-04-1
MDL Number: MFCD03095302
Molecular Formula: C5H3ClFN
Molecular Weight: 131.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 63
Specific Gravity (20/20): 1.35
Formula:
C₅H₃ClFN Boiling Pt: 80 °C Density: 1.35 (20 °C) Flash Pt: 63 °C |
MDL Number:
MFCD03095302 CAS Number: 17282-04-1 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...