2-Chloro-4-fluoro-5-nitrobenzoic acid ≥98.0%

Supplier: TCI America

C2446-5G C2446-25G
TCC2446-5GEA 134.3 CAD
TCC2446-5G TCC2446-25G
2-Chloro-4-fluoro-5-nitrobenzoic acid ≥98.0%
2-Chloro-4-fluoro-5-nitrobenzoic acid

CAS Number: 114776-15-7
MDL Number: MFCD04115715
Molecular Formula: C7H3ClFNO4
Molecular Weight: 219.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 154

Formula: C₇H₃ClFNO
MW: 219.55 g/mol
Melting Pt: 154 °C
MDL Number: MFCD04115715
CAS Number: 114776-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa4be4e811040f5d184948464c232f71458a0a7&vid=6fa4be4e811040f5d184948464c232f71458a0a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR