Cyclopentylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2442-1G
C2442-5G
TCC2442-1GEA
41.08
CAD
TCC2442-1G
TCC2442-5G
Cyclopentylboronic acid (contains varying amounts of Anhydride)
Cyclopentylboronic acid
CAS Number: 63076-51-7
MDL Number: MFCD01074541
Molecular Formula: C5H11BO2
Molecular Weight: 113.95
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 126
Formula:
C₅H₁₁BO₂ Melting Pt: 126 °C |
MDL Number:
MFCD01074541 CAS Number: 63076-51-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...