5-Chloro-2-thiophenesulfonamide ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-5-sulfamoylthiophene, 5-Chlorothiophene-2-sulphonamide, 5-Chlorothiophene-2-sulfonamide, 2-Chloro-5-sulphamoylthiophene

C2441-1G C2441-5G
TCC2441-1GEA 89.27 CAD
TCC2441-1G TCC2441-5G
5-Chloro-2-thiophenesulfonamide ≥98.0%
5-Chloro-2-thiophenesulfonamide

CAS Number: 53595-66-7
MDL Number: MFCD00052584
Molecular Formula: C4H4ClNO2S2
Molecular Weight: 197.65
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 2-Chloro-5-sulfamoylthiophene

Formula: C₄H₄ClNO₂S₂
MW: 197.65 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00052584
CAS Number: 53595-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa450568ea1d9e294882843d641a00694d28021c&vid=aa450568ea1d9e294882843d641a00694d28021c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR