(±)-Cbz-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

C2440-1G C2440-5G
TCC2440-1GEA 83.74 CAD
TCC2440-1G TCC2440-5G
(±)-Cbz-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)
(±)-Cbz-α-phosphonoglycine trimethyl ester

CAS Number: 88568-95-0
MDL Number: MFCD00043304
Molecular Formula: C13H18NO7P
Molecular Weight: 331.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 80


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Cbz-2-phosphonoglycine Trimethyl Ester, Trimethyl 2-(Carbobenzoxyamino)-2-phosphonoacetate, Trimethyl 2-(Cbz-amino)-2-phosphonoacetate

Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00043304
CAS Number: 88568-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb681feb3b3cdee8fd51f33326fffbe99b2382b3&vid=eb681feb3b3cdee8fd51f33326fffbe99b2382b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR