Benzyl-3-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Benzyloxycarbonyl-3-pyrrolidone, 1-Carbobenzoxy-3-pyrrolidone, 1-Cbz-3-pyrrolidinone, 1-Cbz-3-pyrrolidone, Benzyl 3-oxopyrrolidine-1-carboxylate, 1-Benzyloxycarbonyl-3-pyrrolidinone, 1-Carbobenzoxy-3-pyrrolidinone

C2437-1G C2437-5G
TCC2437-1GEA 52.14 CAD
TCC2437-1G TCC2437-5G
Benzyl-3-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
Benzyl-3-oxopyrrolidine-1-carboxylate

CAS Number: 130312-02-6
MDL Number: MFCD03001711
Molecular Formula: C12H13NO3
Molecular Weight: 219.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110


Other Product Names / Synonyms: 1-Cbz-3-pyrrolidinone, 1-Carbobenzoxy-3-pyrrolidinone, 1-Cbz-3-pyrrolidone

Formula: C₁₂H₁₃NO₃
MW: 219.2365 g/mol
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD03001711
CAS Number: 130312-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29736aa0e689989dfd313ba82cd7a548e9401a93&vid=29736aa0e689989dfd313ba82cd7a548e9401a93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR