7-Chloroindole ≥98.0% (by GC)

Supplier: TCI America

C2436-1G C2436-5G
TCC2436-1GEA 101.12 CAD
TCC2436-1G TCC2436-5G
7-Chloroindole ≥98.0% (by GC)
7-Chloroindole

CAS Number: 53924-05-3
MDL Number: MFCD01321301
Molecular Formula: C8H6ClN
Molecular Weight: 151.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 58

Formula: C₈H₆ClN
Boiling Pt: 95 °C
Melting Pt: 58 °C
MDL Number: MFCD01321301
CAS Number: 53924-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35220f55ec4bcb32cb4dcdcbce9985c52fd95061&vid=35220f55ec4bcb32cb4dcdcbce9985c52fd95061&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR